Learn More
Medchemexpress LLC Ethanaminium, 2-hydroxy-N-(iodomethyl)-N,N-dimethyl-, iodide | 28508-22-7 | MFCD20489393 | 98.0% | 356.97 | C5H13I2NO | 250 MG
SDP
Catalog No. 5000283657
Encompass_Preferred
Iodocholine iodide is an iodinated quaternary ammonium reagent used as a metabolizable "green" catalyst for free-radical polymerization and as the non-radioactive iodide analogue of C-11 choline for imaging research. Supplied as a white to off-white solid with high, laboratory-grade purity, it is intended for research applications with recommended storage to preserve stability.
  • Iodinated quaternary ammonium salt suitable for polymerization studies.
  • Non-radioactive analogue of C-11 choline for PET imaging research.
  • High purity (≈98%) for consistent experimental results.
  • Solid, white to off-white appearance for easy handling.
  • Defined storage conditions to maintain stability.
  • Available in multiple pack sizes for flexible ordering.
Catalog No. 50-002-83657 Supplier Medchemexpress LLC Supplier No. HYW130177250MG
Iodocholine iodide is an iodinated quaternary ammonium reagent used as a metabolizable "green" catalyst for free-radical polymerization and as the non-radioactive iodide analogue of C-11 choline for imaging research. Supplied as a white to off-white solid with high, laboratory-grade purity, it is intended for research applications with recommended storage to preserve stability.
  • Iodinated quaternary ammonium salt suitable for polymerization studies.
  • Non-radioactive analogue of C-11 choline for PET imaging research.
  • High purity (≈98%) for consistent experimental results.
  • Solid, white to off-white appearance for easy handling.
  • Defined storage conditions to maintain stability.
  • Available in multiple pack sizes for flexible ordering.