Learn More
Medchemexpress LLC Ethanaminium, 2-hydroxy-N-(iodomethyl)-N,N-dimethyl-, iodide | 28508-22-7 | MFCD20489393 | 98.0% | 356.97 g/mol | C5H13I2NO | 500 MG
SDP
Catalog No. 5000283276
Encompass_Preferred
Iodocholine iodide is a quaternary ammonium iodide used as a non-radioactive analogue of C-11 choline and as a metabolizable catalyst for free-radical polymerization. Supplied as a solid reagent, it is intended for biochemical assays, radiochemistry standards, and polymer chemistry research; analytical documentation (COA, SDS, NMR, LCMS) is available.
  • Non-radioactive standard for choline radiochemistry.
  • Metabolizable catalyst for free-radical polymerization.
  • High purity suitable for analytical and research applications.
  • Available in multiple laboratory pack sizes for flexible use.
  • Accompanied by COA, SDS, NMR, and LCMS documentation.
Catalog No. 50-002-83276 Supplier Medchemexpress LLC Supplier No. HYW130177500MG
Iodocholine iodide is a quaternary ammonium iodide used as a non-radioactive analogue of C-11 choline and as a metabolizable catalyst for free-radical polymerization. Supplied as a solid reagent, it is intended for biochemical assays, radiochemistry standards, and polymer chemistry research; analytical documentation (COA, SDS, NMR, LCMS) is available.
  • Non-radioactive standard for choline radiochemistry.
  • Metabolizable catalyst for free-radical polymerization.
  • High purity suitable for analytical and research applications.
  • Available in multiple laboratory pack sizes for flexible use.
  • Accompanied by COA, SDS, NMR, and LCMS documentation.