Learn More
Medchemexpress LLC Ethanaminium, 2-hydroxy-N,N,N-trimethyl-, chloride (1:1)-13C3 | 67-48-1 | 99.7% | 5 MG
SDP
Catalog No. 5000411998
Encompass_Preferred
Choline Chloride-13C3 is the 13C-labeled Choline (chloride). It is an organic compound and a quaternary ammonium salt, an acyl group acceptor and choline acetyltransferase substrate. It is also an important additive in feed, especially for chickens, where it accelerates growth. For research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-11998 Supplier Medchemexpress LLC Supplier No. HYB1337S45MG
Choline Chloride-13C3 is the 13C-labeled Choline (chloride). It is an organic compound and a quaternary ammonium salt, an acyl group acceptor and choline acetyltransferase substrate. It is also an important additive in feed, especially for chickens, where it accelerates growth. For research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS