Learn More
Medchemexpress LLC Ethanaminium, N-(carboxymethyl)-2-([3R]-1-[8-([4-(1,1-dimethylethyl)-2-thiazolyl]amino)...) inner salt | 957471-96-4 | 99.9% | 693.78 | C31H39N11O6S | 100MG
SDP
Catalog No. 5000239300
Encompass_Preferred
D13-9001 is a small-molecule bacterial efflux pump inhibitor for biochemical and microbiology research into efflux-mediated antibiotic resistance. It selectively targets RND family efflux pump subunits AcrB and MexB, with reported binding affinities (KD) of 1.15 μM (AcrB) and 3.57 μM (MexB). The compound is provided as a high-purity solid suitable for preparing stock solutions and in-vitro assays.
  • Potent inhibitor of AcrB and MexB efflux pump subunits.
  • Demonstrates KD = 1.15 μM (AcrB) and KD = 3.57 μM (MexB).
  • Exhibits antibacterial activity in vitro.
  • High purity suitable for biochemical assays.
  • Supplied as a solid for stock solution preparation.
  • Available in multiple pack sizes including 100 mg.
Catalog No. 50-002-39300 Supplier Medchemexpress LLC Supplier No. HY124819100MG
Add to Cart
Add to Cart
Request a Quote
D13-9001 is a small-molecule bacterial efflux pump inhibitor for biochemical and microbiology research into efflux-mediated antibiotic resistance. It selectively targets RND family efflux pump subunits AcrB and MexB, with reported binding affinities (KD) of 1.15 μM (AcrB) and 3.57 μM (MexB). The compound is provided as a high-purity solid suitable for preparing stock solutions and in-vitro assays.
  • Potent inhibitor of AcrB and MexB efflux pump subunits.
  • Demonstrates KD = 1.15 μM (AcrB) and KD = 3.57 μM (MexB).
  • Exhibits antibacterial activity in vitro.
  • High purity suitable for biochemical assays.
  • Supplied as a solid for stock solution preparation.
  • Available in multiple pack sizes including 100 mg.