Learn More
Medchemexpress LLC Ethanaminium, N,N,N-trimethyl-2-(phosphonooxy)-, chloride, sodium salt (1:1:2) | 16904-96-4 | MFCD00070597 | 98.0% | 263.57 g/mol | C5H13ClNNa2O4P | 500 MG
SDP
Catalog No. 5000283096
Encompass_Preferred
Phosphocholine chloride disodium is the disodium salt form of phosphocholine used as a biochemical reagent for preparing aqueous solutions and studying phospholipid headgroup behavior, membrane models, and drug-delivery research. It is supplied as a solid with documented solubility, assay, and batch data to support reproducible experimental preparation.
  • Biochemical reagent for membrane and lipid research.
  • Supplied as a solid for preparing aqueous solutions.
  • Documented solubility data for accurate molarity calculations.
  • Batch certificate of analysis available to support quality control.
  • Suitable for preparation of defined experimental concentrations.
Catalog No. 50-002-83096 Supplier Medchemexpress LLC Supplier No. HYW127640500MG
Phosphocholine chloride disodium is the disodium salt form of phosphocholine used as a biochemical reagent for preparing aqueous solutions and studying phospholipid headgroup behavior, membrane models, and drug-delivery research. It is supplied as a solid with documented solubility, assay, and batch data to support reproducible experimental preparation.
  • Biochemical reagent for membrane and lipid research.
  • Supplied as a solid for preparing aqueous solutions.
  • Documented solubility data for accurate molarity calculations.
  • Batch certificate of analysis available to support quality control.
  • Suitable for preparation of defined experimental concentrations.