Learn More
Medchemexpress LLC Ethanaminium, N,N,N-trimethyl-2-(phosphonooxy)-, chloride, sodium salt | 16904-96-4 | MFCD09037359 | 98.0% | 263.57 g·mol⁻¹ | C5H13ClNNa2O4P | 250 MG
SDP
Catalog No. 5000283517
Encompass_Preferred
Phosphocholine (chloride disodium) is the disodium salt of phosphocholine supplied as a white to off-white solid for life-science research. It functions as a biochemical reagent and formulation additive for drug-delivery and membrane studies; analytical data (1H NMR) and the certificate of analysis report identity and 98.0% purity. CAS 16904-96-4; molecular weight 263.57 g·mol⁻¹; formula C5H13ClNNa2O4P. Store sealed, away from moisture, at 4 °C; in solvent store at -80 °C (6 months) or -20 °C (1 month).
  • High purity for analytical and research use.
  • White to off-white solid convenient for handling and formulation.
  • Identity confirmed by 1H NMR.
  • Suitable for use in drug-delivery and membrane studies.
  • Stable when stored sealed and protected from moisture.
Catalog No. 50-002-83517 Supplier Medchemexpress LLC Supplier No. HYW127640250MG
Phosphocholine (chloride disodium) is the disodium salt of phosphocholine supplied as a white to off-white solid for life-science research. It functions as a biochemical reagent and formulation additive for drug-delivery and membrane studies; analytical data (1H NMR) and the certificate of analysis report identity and 98.0% purity. CAS 16904-96-4; molecular weight 263.57 g·mol⁻¹; formula C5H13ClNNa2O4P. Store sealed, away from moisture, at 4 °C; in solvent store at -80 °C (6 months) or -20 °C (1 month).
  • High purity for analytical and research use.
  • White to off-white solid convenient for handling and formulation.
  • Identity confirmed by 1H NMR.
  • Suitable for use in drug-delivery and membrane studies.
  • Stable when stored sealed and protected from moisture.