Learn More
Medchemexpress LLC Ethane-1,2-diamine-d4 dihydrochloride | 34334-71-9 | 137.04 | 500 MG
SDP
Catalog No. 5000365688
Encompass_Preferred
Ethane-1,2-diamine-d4 (Ethylenediamine-d4) dihydrochloride is a deuterium labeled Ethane-1,2-diamine. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuterium labeled compound
  • Incorporated into drug molecules as tracers for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-65688 Supplier Medchemexpress LLC Supplier No. 500MGHYW012684S1
Ethane-1,2-diamine-d4 (Ethylenediamine-d4) dihydrochloride is a deuterium labeled Ethane-1,2-diamine. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuterium labeled compound
  • Incorporated into drug molecules as tracers for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs