Learn More
Medchemexpress LLC Ethane-1,2-diamine-d4 dihydrochloride | 34334-71-9 | MFCD00064506 | 137.04 | 100 MG
SDP
Catalog No. 5000365003
Encompass_Preferred
Ethane-1,2-diamine-d4 dihydrochloride (Ethylenediamine-d4) is a deuterium labeled Ethane-1,2-diamine. Stable heavy isotopes have been incorporated into drug molecules as tracers for quantitation during drug development, with deuteration potentially affecting pharmacokinetic and metabolic profiles.
  • Isotope-labeled compounds
  • Store at room temperature, keep dry and cool
  • In solvent: -80°C for 1 year, -20°C for 6 months
Catalog No. 50-003-65003 Supplier Medchemexpress LLC Supplier No. 100MGHYW012684S1
Ethane-1,2-diamine-d4 dihydrochloride (Ethylenediamine-d4) is a deuterium labeled Ethane-1,2-diamine. Stable heavy isotopes have been incorporated into drug molecules as tracers for quantitation during drug development, with deuteration potentially affecting pharmacokinetic and metabolic profiles.
  • Isotope-labeled compounds
  • Store at room temperature, keep dry and cool
  • In solvent: -80°C for 1 year, -20°C for 6 months