Learn More
Mitegen Ethane Cup for Vitrobot Dewar By Nanosoft
SDP
Catalog No. 5000495034
Encompass
The Nanosoft Ethane Cup is a brass cup with the same dimensions as the one found in the Thermo Fisher Vitrobot dewar set. This design ensures it works seamlessly with the styrofoam dewar set from the Vitrobot, providing equal performance and a reliable option for cryo-EM sample vitrification. This product is a crucial component for the cryo-vitrification process, ensuring a smooth and efficient workflow.
Catalog No. 50-004-95034 Supplier Mitegen Supplier No. MCEMNSECUP01
The Nanosoft Ethane Cup is a brass cup with the same dimensions as the one found in the Thermo Fisher Vitrobot dewar set. This design ensures it works seamlessly with the styrofoam dewar set from the Vitrobot, providing equal performance and a reliable option for cryo-EM sample vitrification. This product is a crucial component for the cryo-vitrification process, ensuring a smooth and efficient workflow.