Learn More
Mitegen Ethane-Temperature Controlled Dewar By Nanosoft For Vitrobot, Precise Cryo-EM Plunge Freezing, Integrated Tool Warmer, User-Friendly OLED Control
SDP
Catalog No. NC3105761
Encompass
This Ethane-Temperature Controlled Vitrobot Dewar by Nanosoft enables highly accurate and precise control of liquid ethane temperature during critical plunge freezing in cryo-electron microscopy (cryo-EM). Embedded heaters and sensors within the brass ethane cup ensure consistent temperatures, even away from the ethane's melting point, enhancing reliability and consistency in sample preparation. A unique "Snowflake Setting" maintains the ethane just above freezing, creating a thin crust of frozen ethane for optimized conditions. The integrated control box features a warming plate and air flow, allowing quick drying of cryo-tools. This robust system includes a high-quality foam dewar and a cryogenically-compatible plastic basin, both easily disassembled without tools for efficient drying via a built-in bake-out cycle. Its user-friendly interface, with an OLED screen and intuitive knob, streamlines operation, making precise cryo-EM sample vitrification more dependable and accessible.
Catalog No. NC3105761 Supplier Mitegen Supplier No. MCEMNSETCVBD
Add to Cart
Add to Cart
Request a Quote
This Ethane-Temperature Controlled Vitrobot Dewar by Nanosoft enables highly accurate and precise control of liquid ethane temperature during critical plunge freezing in cryo-electron microscopy (cryo-EM). Embedded heaters and sensors within the brass ethane cup ensure consistent temperatures, even away from the ethane's melting point, enhancing reliability and consistency in sample preparation. A unique "Snowflake Setting" maintains the ethane just above freezing, creating a thin crust of frozen ethane for optimized conditions. The integrated control box features a warming plate and air flow, allowing quick drying of cryo-tools. This robust system includes a high-quality foam dewar and a cryogenically-compatible plastic basin, both easily disassembled without tools for efficient drying via a built-in bake-out cycle. Its user-friendly interface, with an OLED screen and intuitive knob, streamlines operation, making precise cryo-EM sample vitrification more dependable and accessible.