Learn More
Medchemexpress LLC Ethanedioic acid, disodium salt | 62-76-0 | MFCD00012465 | 99.7% | 134.00 g/mol | Na2C2O4 | 1g
SDP
Catalog No. 5000267885
Encompass_Preferred
Sodium oxalate is an orally active dispersant and coordination agent Sodium oxalate causes mitochondrial dysfunction Sodium oxalate has catalytic enhancing activity Sodium oxalate induces stable chronic kidney disease Sodium oxalate induces highly malignant and undifferentiated breast tumors[1][2][3][4][5][6][7][8]
Catalog No. 50-002-67885 Supplier Medchemexpress LLC Supplier No. HYY0262B1G
Sodium oxalate is an orally active dispersant and coordination agent Sodium oxalate causes mitochondrial dysfunction Sodium oxalate has catalytic enhancing activity Sodium oxalate induces stable chronic kidney disease Sodium oxalate induces highly malignant and undifferentiated breast tumors[1][2][3][4][5][6][7][8]