Learn More
AdipoGen Ethanediyl bis-1H-triazole
SDP
Catalog No. 502560978
Encompass_Preferred
Chemical. CAS 116476-98-3. Formula C6H8N6. MW 164.17. Synthetic. Building block for synthesis.
Catalog No. 50-256-0978 Supplier AdipoGen Supplier No. CDXE0066G001
Chemical. CAS 116476-98-3. Formula C6H8N6. MW 164.17. Synthetic. Building block for synthesis.