Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethanesulfonic acid, 2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]- | 1817735-43-5 | 99.5% | 257.30 | 25 MG
SDP

Catalog No. 5000349053
Encompass_Preferred

Amino-PEG3-C2-sulfonic acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker, one binding to an E3 ubiquitin ligase and the other to a target protein. They function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Intended for research use only
  • Appearance: oil, colorless to light yellow
  • Target: PEGs
  • Storage: -20°C under nitrogen

Catalog No. 50-003-49053 Supplier Medchemexpress LLC Supplier No. 25MGHY140001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amino-PEG3-C2-sulfonic acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker, one binding to an E3 ubiquitin ligase and the other to a target protein. They function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Intended for research use only
  • Appearance: oil, colorless to light yellow
  • Target: PEGs
  • Storage: -20°C under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.