Learn More
Medchemexpress LLC Ethanesulfonic acid, 2-[[(3α,5β,7α,12α)-3,7,12-trihydroxy-24-oxocholan-24-yl-2,2,4,4-d4]amino]- | 252030-90-3 | 99.5% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB1788S
Encompass_Preferred
Taurocholic acid-d4 is deuterium labeled Taurocholic acid, a bile acid that participates in the emulsification of fats. Deuterium substitution in drug molecules can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-14219