Learn More
Medchemexpress LLC Ethanesulfonic acid, 2-[[(3α,5β,7α,12α)-3,7,12-trihydroxy-24-oxocholan-24-yl-2,2,4,4-d4]amino]- | 252030-90-3 | 99.5% | 5 MG
SDP
Catalog No. 5000412729
Encompass_Preferred
Taurocholic acid-d4 is a deuterium-labeled form of Taurocholic acid, which is a bile acid crucial for the emulsification of fats. This product can be utilized as a tracer in various applications or as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-12729 Supplier Medchemexpress LLC Supplier No. HYB1788S5MG
Taurocholic acid-d4 is a deuterium-labeled form of Taurocholic acid, which is a bile acid crucial for the emulsification of fats. This product can be utilized as a tracer in various applications or as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules
  • Potential to affect pharmacokinetic and metabolic profiles of drugs