Learn More
Medchemexpress LLC Ethanesulfonic acid, 2-[(aminoiminomethyl)amino]- | 543-18-0 | 99.9% | 167.19 | 100 MG
SDP
Catalog No. 5000436072
Encompass_Preferred
Guanidinoethyl sulfonate (Standard) is an analytical standard and an orally available, blood-brain permeable competitive inhibitor of taurine transporters. It acts as a competitive antagonist of glycine receptors (GlyR) (IC50=565 μM) and shows weak agonist/antagonist effects on GABAA receptors. It inhibits taurine transmembrane transport, blocks glycine-mediated chloride influx, and may regulate brain pH for neuroprotective effects, useful in studies of ischemic brain injury. It is applied in qualitative, quantitative, and methodological research experiments including HPLC, GC, and MS.
  • Analytical standard for research
  • Orally available and blood-brain permeable
  • Inhibits taurine transporters
  • Antagonizes glycine receptors
  • Modulates GABAA receptors
  • Blocks glycine-mediated chloride influx
  • May offer neuroprotective effects
  • Used in HPLC, GC, and MS applications
Catalog No. 50-004-36072 Supplier Medchemexpress LLC Supplier No. HY113329R100MG
Guanidinoethyl sulfonate (Standard) is an analytical standard and an orally available, blood-brain permeable competitive inhibitor of taurine transporters. It acts as a competitive antagonist of glycine receptors (GlyR) (IC50=565 μM) and shows weak agonist/antagonist effects on GABAA receptors. It inhibits taurine transmembrane transport, blocks glycine-mediated chloride influx, and may regulate brain pH for neuroprotective effects, useful in studies of ischemic brain injury. It is applied in qualitative, quantitative, and methodological research experiments including HPLC, GC, and MS.
  • Analytical standard for research
  • Orally available and blood-brain permeable
  • Inhibits taurine transporters
  • Antagonizes glycine receptors
  • Modulates GABAA receptors
  • Blocks glycine-mediated chloride influx
  • May offer neuroprotective effects
  • Used in HPLC, GC, and MS applications