Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethanesulfonic acid, 2-bromo-, sodium salt | 4263-52-9 | MFCD00007530 | 97.0% | 211.01 | C2H4BrNaO3S | 50 G
SDP

Catalog No. 5000303992
Encompass_Preferred

Sodium 2-bromoethanesulfonate is the sodium salt of 2-bromoethanesulfonic acid supplied as a white to off-white solid reagent for life-science and chemical research. It is used as a biochemical reagent and research chemical where a brominated ethanesulfonate derivative is required; data sheet, SDS, and COA are available for specification and safety information.

  • Solid, white to off-white appearance.
  • Chemical formula C2H4BrNaO3S; molecular weight 211.01.
  • CAS number 4263-52-9.
  • Typical purity 97.0% (assay documentation may report 99.7%).
  • Available in multiple package sizes including 25 g, 50 g, 100 g, 250 g, and 500 g.
  • Data sheet, COA, and SDS available for batch and safety details.

Catalog No. 50-003-03992 Supplier Medchemexpress LLC Supplier No. HYW01435550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium 2-bromoethanesulfonate is the sodium salt of 2-bromoethanesulfonic acid supplied as a white to off-white solid reagent for life-science and chemical research. It is used as a biochemical reagent and research chemical where a brominated ethanesulfonate derivative is required; data sheet, SDS, and COA are available for specification and safety information.

  • Solid, white to off-white appearance.
  • Chemical formula C2H4BrNaO3S; molecular weight 211.01.
  • CAS number 4263-52-9.
  • Typical purity 97.0% (assay documentation may report 99.7%).
  • Available in multiple package sizes including 25 g, 50 g, 100 g, 250 g, and 500 g.
  • Data sheet, COA, and SDS available for batch and safety details.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.