Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pfaltz & Bauer ETHANESULFONIC ACID 97%

Catalog No. 507490840
Encompass

Ethanesulfonic Acid 97%; 10g; CAS: 594-45-6

Catalog No. 50-749-0840 Supplier Pfaltz & Bauer Supplier No. E0265010G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethanesulfonic Acid 97%; 10g; CAS: 594-45-6

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.