Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanesulfonyl chloride, 98+%, ACROS Organics™

Click to view available options
Quantity:
5mL
50mL
100mL
500mL
2.5L
Packaging:
Glass bottle
594-44-5
C2H5ClO2S
128.58
MFCD00007460
FRYHCSODNHYDPU-UHFFFAOYSA-N
ethylsulfonyl chloride,ethylsulfochloride,1-ethanesulfonyl chloride,ethanesulphonyl chloride,ethanesulfonylchloride,unii-b56zoc898a,ethylsulfonylchloride,ethansulfonyl chloride,ethansulfonylchlorid,ethanesulfonylchoride
11667
ethanesulfonyl chloride
CCS(=O)(=O)Cl
594-44-5
C2H5ClO2S
128.58
MFCD00007460
FRYHCSODNHYDPU-UHFFFAOYSA-N
ethylsulfonyl chloride,ethylsulfochloride,1-ethanesulfonyl chloride,ethanesulphonyl chloride,ethanesulfonylchloride,unii-b56zoc898a,ethylsulfonylchloride,ethansulfonyl chloride,ethansulfonylchlorid,ethanesulfonylchoride
11667
ethanesulfonyl chloride
CCS(=O)(=O)Cl
Viscosity 1.44 mm2/s (20°C)
Refractive Index 1.4520 to 1.4540
Density 1.3500g/mL
Quantity 500mL
Infrared Spectrum Authentic
Flash Point 83°C
Boiling Point 177.0°C
Packaging Glass bottle
Color Brown to Pink-Purple or Colorless to Yellow
Specific Gravity 1.35
Assay Percent Range 98% min. (GC)
Linear Formula C2H5SO2Cl
Beilstein 04, 6
Solubility Information Solubility in water: decomposes in water. Other solubilities: soluble in acetone and ethyl ether
Formula Weight 128.58
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material Ethanesulfonyl chloride, 98+%
Show More Show Less

GHS H Statement:
Toxic if inhaled.
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-842-5

RTECSNumber : KI8050000

TSCA : TSCA

Recommended Storage : this product darkens during storage