Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethanethioic acid S-[2-[6-[[[4-[3-(dimethylamino)propoxy]phenyl]sulfonyl]amino]-3-pyridinyl]-2-oxoet | 940943-37-3 | 98.1% | 451.56 | C20H25N3O5S2 | 5 MG
SDP

Catalog No. 5000241455
Encompass_Preferred

KD 5170 is a mercaptoketone-based pan inhibitor of histone deacetylases (HDACs) that exhibits broad-spectrum antitumor activity in vitro and in vivo. It is supplied as a research compound with characterized purity and physicochemical properties for preclinical studies.

  • Pan inhibitor of histone deacetylases (HDACs).
  • Demonstrated broad-spectrum antitumor activity in vitro and in vivo.
  • High purity suitable for research applications (≈98.1%).
  • Soluble in DMSO at high concentration (100 mg/mL) for in vitro assays.
  • Recommended solid storage at 4°C; solutions stable at -80°C (6 months) or -20°C (1 month).
  • Molecular formula C20H25N3O5S2 and molecular weight 451.56 aid compound handling and dosing.

Catalog No. 50-002-41455 Supplier Medchemexpress LLC Supplier No. HY1075495MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KD 5170 is a mercaptoketone-based pan inhibitor of histone deacetylases (HDACs) that exhibits broad-spectrum antitumor activity in vitro and in vivo. It is supplied as a research compound with characterized purity and physicochemical properties for preclinical studies.

  • Pan inhibitor of histone deacetylases (HDACs).
  • Demonstrated broad-spectrum antitumor activity in vitro and in vivo.
  • High purity suitable for research applications (≈98.1%).
  • Soluble in DMSO at high concentration (100 mg/mL) for in vitro assays.
  • Recommended solid storage at 4°C; solutions stable at -80°C (6 months) or -20°C (1 month).
  • Molecular formula C20H25N3O5S2 and molecular weight 451.56 aid compound handling and dosing.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.