Learn More
Medchemexpress LLC Ethanethiol, 2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]- | 1189760-69-7 | 97.0% | 50 MG
SDP
Catalog No. 5000340346
Encompass_Preferred
HS-PEG3-CH2CH2NH2 is a PEG-based PROTAC linker, also known as Thiol-PEG3-amine, utilized in the synthesis of PROTACs. While the salt and free forms exhibit comparable biological activity, the hydrochloride salt typically offers enhanced water solubility and stability.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-40346 Supplier Medchemexpress LLC Supplier No. HY130871A50MG
HS-PEG3-CH2CH2NH2 is a PEG-based PROTAC linker, also known as Thiol-PEG3-amine, utilized in the synthesis of PROTACs. While the salt and free forms exhibit comparable biological activity, the hydrochloride salt typically offers enhanced water solubility and stability.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins