Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Benzoate, 99+%

Catalog No. AC118090025
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC118090025 2.5 kg Glass bottle
AC118095000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC118090025 Supplier Thermo Scientific Chemicals Supplier No. 118090025
Only null left
Add to Cart

CAS: 93-89-0 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-89-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00009109
InChI Key MTZQAGJQAFMTAQ-UHFFFAOYSA-N
Synonym benzoic acid, ethyl ester, benzoic ether, ethyl benzenecarboxylate, ethylbenzoate, benzoyl ethyl ether, benzoic acid ethyl ester, ethylester kyseliny benzoove, natural, unii-j115brj15h, fema no. 2422
PubChem CID 7165
IUPAC Name ethyl benzoate
SMILES CCOC(=O)C1=CC=CC=C1

Specifications

Acidity 1mg KOH/g max.
Melting Point -34.0°C
Color Colorless to Yellow
Density 1.0450g/mL
Boiling Point 212.0°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5040 to 1.5060
Linear Formula C6H5CO2C2H5
Quantity 2.5 kg
Beilstein 09, IV, 285
Specific Gravity 1.045
Merck Index 15, 3821
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and chloroform
Formula Weight 150.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Ethyl benzoate, 99+%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-284-3
RTECSNumber DH0200000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.