Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, 140 proof (70%), Decon™ Labs
GREENER_CHOICE

Catalog No. 04355350 Shop All Decon Labs Products
Change view
Click to view available options
Quantity:
32 oz.
5 gal.
4 x 1 gal.
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
04-355-350 5 gal. Plastic Bottle
04-355-309 32 oz. Plastic Bottle
04-355-305 4 x 1 gal. Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 04-355-350 Supplier Decon Laboratories, Inc. Supplier No. 2405
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Note: May require a federal tax exempt certificate. For more information regarding Ethyl Alcohol regulations, contact Fisher Customer Service.

Product 04-355-305: May require a federal tax exempt certificate.Service teams.

Chemical Identifiers

CAS 64-17-5 , 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07 g/mol
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name Ethanol
SMILES CCO

Specifications

Melting Point -114°C
CAS Min % 62%
Color Clear
CAS Max % 70%
Density 7.109
Quantity 5 gal.
Packaging Plastic Bottle
Physical Form Liquid
UN Number UN1170
Boiling Point 78.3°C
Percent Purity 70%
Grade USP
Chemical Name or Material Ethanol, 140 Proof (70%)
Show More Show Less
DOTInformation DOT Class: 3, Packaging: II
Recommended Storage Do not store alcohol above 120°F
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.