Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, 140 proof (70%), Tax-Paid, Decon™ Labs
GREENER_CHOICE

Catalog No. 07678001 Shop All Decon Labs Products
Change view
Click to view available options
Quantity:
5 gal.
4 x 1 gal.
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
07-678-001 4 x 1 gal. Plastic Bottle
04-355-351 5 gal. Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-678-001 Supplier Decon Laboratories, Inc. Supplier No. 2401TP
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Federal Tax is included in the price

Product 07-678-001: May require a federal tax exempt certificate.

Chemical Identifiers

CAS 64-17-5 , 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07 g/mol
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name Ethanol
SMILES CCO

Specifications

Melting Point -114°C
Color Clear
Density 7.11 lb./gal.
Quantity 4 x 1 gal.
Packaging Plastic Bottle
Physical Form Liquid
UN Number UN1170
Boiling Point 78.3°C
Percent Purity 70%
Grade USP
Chemical Name or Material Ethanol, 140 Proof (70%)
Show More Show Less
DOTInformation DOT Class: 3, Packaging: II
Recommended Storage Do not store alcohol above 120°F
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.