Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, 190 Proof USP (Ethyl Alcohol, 95%), Reagents

Catalog No. 01338164
Change view
Click to view available options
Quantity:
55 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-338-164 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-338-164 Supplier Reagents Holdings Llc Supplier No. C111190055E
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethanol, 190 Proof USP (Ethyl Alcohol, 95%), Meets USP specifications, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Ethanol, Ethyl alcohol, Methylcarbinol
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -112°C
Color Colorless
Quantity 55 gal.
Packaging ETHANOL USP 190 PROOF TNI
Physical Form Liquid
Specific Gravity 0.816
Boiling Point 79°C
Percent Purity 94.9 to 96.0%
Chemical Name or Material Ethanol, 190 Proof USP (Ethyl Alcohol, 95%)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.