Learn More
Medchemexpress LLC Ethanol, 2-[2-[2-[2-[(2,4-dinitrophenyl)amino]ethoxy]ethoxy]ethoxy]- | 1807520-99-5 | 97.9% | C14H21N3O8 | 250 MG
SDP
Catalog No. 5000352887
Encompass_Preferred
DNP-PEG4-alcohol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds that contain two different ligands connected by a linker; one targets an E3 ubiquitin ligase and the other targets a protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Facilitates selective degradation of target proteins.
Catalog No. 50-003-52887 Supplier Medchemexpress LLC Supplier No. 250MGHY140618
DNP-PEG4-alcohol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds that contain two different ligands connected by a linker; one targets an E3 ubiquitin ligase and the other targets a protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Facilitates selective degradation of target proteins.