Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethanol, 2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]- | 86770-67-4 | 98.88% | 219.24 | 250 MG
SDP

Catalog No. 5000402648
Encompass_Preferred

Azido-PEG4-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is a click chemistry reagent containing an Azide group, which can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.

  • Peg-based PROTAC linker
  • Click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Appearance: liquid
  • Color: colorless to light yellow

Catalog No. 50-004-02648 Supplier Medchemexpress LLC Supplier No. 250MGHY22340
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azido-PEG4-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is a click chemistry reagent containing an Azide group, which can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.

  • Peg-based PROTAC linker
  • Click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Appearance: liquid
  • Color: colorless to light yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.