Learn More
Medchemexpress LLC Ethanol, 2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]- | 86770-67-4 | 98.9% | 219.24 | 10 G
SDP
Catalog No. 5000401155
Encompass_Preferred
Azido-PEG4-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. It acts as a click chemistry reagent, featuring an azide group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules. It also facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs employ two ligands linked by a linker-one for an E3 ubiquitin ligase and another for the target protein-to selectively degrade target proteins via the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-004-01155 Supplier Medchemexpress LLC Supplier No. HY2234010G
Azido-PEG4-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. It acts as a click chemistry reagent, featuring an azide group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules. It also facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs employ two ligands linked by a linker-one for an E3 ubiquitin ligase and another for the target protein-to selectively degrade target proteins via the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions