Learn More
Medchemexpress LLC Ethanol, 2-[2-[2-(2-mercaptoethoxy)ethoxy]ethoxy]- | 90952-27-5 | 99.9% | 210.29 | 1 G
SDP
Catalog No. 5000365321
Encompass_Preferred
Thiol-PEG4-alcohol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These molecules connect a ligand for an E3 ubiquitin ligase with a target protein ligand, enabling the selective degradation of target proteins through the intracellular ubiquitin-proteasome system. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Exploits the ubiquitin-proteasome system
  • For research use only
Catalog No. 50-003-65321 Supplier Medchemexpress LLC Supplier No. 1GHY133292
Thiol-PEG4-alcohol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These molecules connect a ligand for an E3 ubiquitin ligase with a target protein ligand, enabling the selective degradation of target proteins through the intracellular ubiquitin-proteasome system. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Exploits the ubiquitin-proteasome system
  • For research use only