Learn More
Medchemexpress LLC Ethanol, 2-[2-(2-aminoethoxy)ethoxy]- | 6338-55-2 | 149.19 | 10 G
SDP
Catalog No. 5000446591
Encompass_Preferred
NH2-PEG3, also known as PROTAC Linker 35, is a polyethylene glycol (PEG) linker used in the synthesis of PROTACs, including PROTAC (β-NF-JQ1). PROTACs are compounds designed to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Polyethylene glycol (PEG) linker
  • Used in the synthesis of PROTACs
  • Facilitates the synthesis of PROTAC (β-NF-JQ1)
  • Enables targeted protein degradation via the ubiquitin-proteasome system
Catalog No. 50-004-46591 Supplier Medchemexpress LLC Supplier No. HYW00754510G
NH2-PEG3, also known as PROTAC Linker 35, is a polyethylene glycol (PEG) linker used in the synthesis of PROTACs, including PROTAC (β-NF-JQ1). PROTACs are compounds designed to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Polyethylene glycol (PEG) linker
  • Used in the synthesis of PROTACs
  • Facilitates the synthesis of PROTAC (β-NF-JQ1)
  • Enables targeted protein degradation via the ubiquitin-proteasome system