Learn More
Medchemexpress LLC Ethanol, 2-[2-(2-hydroxyethoxy)ethoxy]-, 1-(4-methylbenzenesulfonate) | 77544-68-4 | 97.0% | 304.36 | 25 G
SDP
Catalog No. 5000403297
Encompass_Preferred
Tos-PEG3 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It can also be used for the synthesis of 3'-aminooxy oligonucleotides solid supports.
  • Peg-based protac linker in the synthesis of protacs.
  • Used for the synthesis of 3'-aminooxy oligonucleotides solid supports.
Catalog No. 50-004-03297 Supplier Medchemexpress LLC Supplier No. 25GHY23408
Tos-PEG3 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It can also be used for the synthesis of 3'-aminooxy oligonucleotides solid supports.
  • Peg-based protac linker in the synthesis of protacs.
  • Used for the synthesis of 3'-aminooxy oligonucleotides solid supports.