Learn More
Medchemexpress LLC Ethanol, 2-[2-[2-(methylamino)ethoxy]ethoxy]- | 282551-10-4 | ≥98.0% | 163.21 | 250 MG
SDP
Catalog No. 5000351567
Encompass_Preferred
PEG3-methylamine is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Targets an E3 ubiquitin ligase and a protein of interest
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-51567 Supplier Medchemexpress LLC Supplier No. 250MGHY140161
PEG3-methylamine is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Targets an E3 ubiquitin ligase and a protein of interest
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins