Learn More
Medchemexpress LLC Ethanol, 2-[2-(2-propyn-1-yloxy)ethoxy]-, 1-(4-methylbenzenesulfonate) | 1119249-30-7 | 98.2% | 1 G
SDP
Catalog No. 5000338513
Encompass_Preferred
Tos-PEG2-O-Propargyl is a PEG-based PROTAC linker that functions as a click chemistry reagent. It incorporates an alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. This linker can be utilized in the synthesis of molecules like Thalidomide-O-PEG2-propargyl.
  • Utilized in the synthesis of molecules like Thalidomide-O-PEG2-propargyl.
  • Functions as a click chemistry reagent.
  • Incorporates an alkyne group.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups.
  • A component in PROTACs, which exploit the ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-003-38513 Supplier Medchemexpress LLC Supplier No. HY1301621G
Tos-PEG2-O-Propargyl is a PEG-based PROTAC linker that functions as a click chemistry reagent. It incorporates an alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. This linker can be utilized in the synthesis of molecules like Thalidomide-O-PEG2-propargyl.
  • Utilized in the synthesis of molecules like Thalidomide-O-PEG2-propargyl.
  • Functions as a click chemistry reagent.
  • Incorporates an alkyne group.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups.
  • A component in PROTACs, which exploit the ubiquitin-proteasome system to selectively degrade target proteins.