Learn More
Medchemexpress LLC Ethanol, 2-[2-(2-propyn-1-yloxy)ethoxy]-, 1-methanesulfonate | 943726-01-0 | 95.0% | 1 G
SDP
Catalog No. 5000340215
Encompass_Preferred
Propargyl-PEG2-MS is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting an E3 ubiquitin ligase ligand with a target protein ligand via a linker.
  • PEG-based PROTAC linker
  • Functions as a click chemistry reagent
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Appears as a colorless to light yellow liquid
Catalog No. 50-003-40215 Supplier Medchemexpress LLC Supplier No. HY1305841G
Propargyl-PEG2-MS is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting an E3 ubiquitin ligase ligand with a target protein ligand via a linker.
  • PEG-based PROTAC linker
  • Functions as a click chemistry reagent
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Appears as a colorless to light yellow liquid