Learn More
Medchemexpress LLC Ethanol, 2-[2-(2-propyn-1-yloxy)ethoxy]- | 7218-43-1 | 99.7% | 1 G
SDP
Catalog No. 5000339970
Encompass_Preferred
Propargyl-PEG2-OH is a PEG-based PROTAC linker. It functions as a click chemistry reagent, possessing an alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. This compound can be utilized in the synthesis of Thalidomide-O-PEG2-propargyl.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Utilized in the synthesis of thalidomide-O-PEG2-propargyl
Catalog No. 50-003-39970 Supplier Medchemexpress LLC Supplier No. HY1305411G
Propargyl-PEG2-OH is a PEG-based PROTAC linker. It functions as a click chemistry reagent, possessing an alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. This compound can be utilized in the synthesis of Thalidomide-O-PEG2-propargyl.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Utilized in the synthesis of thalidomide-O-PEG2-propargyl