Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethanol, 2-[2-[2-[(tetrahydro-2H-pyran-2-yl)oxy]ethoxy]ethoxy]- | 60221-37-6 | 98.7% | 25 G
SDP

Catalog No. 5000341780
Encompass_Preferred

THP-PEG3-OH is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs feature two ligands connected by a linker, one for an E3 ubiquitin ligase and another for a target protein. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade specific target proteins.

  • High purity of 98.7%
  • Molecular weight of 234.29
  • Chemical formula is C11H22O5
  • Appears as a liquid
  • Colorless to light yellow
  • Utilizes the ubiquitin-proteasome system
  • Selectively degrades target proteins

Catalog No. 50-003-41780 Supplier Medchemexpress LLC Supplier No. 25GHY130216
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

THP-PEG3-OH is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs feature two ligands connected by a linker, one for an E3 ubiquitin ligase and another for a target protein. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade specific target proteins.

  • High purity of 98.7%
  • Molecular weight of 234.29
  • Chemical formula is C11H22O5
  • Appears as a liquid
  • Colorless to light yellow
  • Utilizes the ubiquitin-proteasome system
  • Selectively degrades target proteins

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.