Learn More
Medchemexpress LLC Ethanol, 2,2'-[[6-[[(4-methoxyphenyl)methyl)amino]-9-(1-methylethyl)-9H-purin-2-yl]imino]bis | 199986-75-9 | C20H28N6O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY15339
Encompass_Preferred
CVT-313 (Cdk2 Inhibitor III) is a potent, selective, reversible, and ATP-competitive inhibitor of CDK2 with an IC50 of 0.5 μM. It effectively inhibits CDC5L phosphorylation and shows profound effects on cell proliferation at concentrations of 5-20 μM. It exhibits high selectivity, requiring much higher concentrations to inhibit other kinases like CDK1 and CDK4.
  • Potent, selective, reversible, and ATP-competitive inhibitor of CDK2
  • Inhibits CDC5L phosphorylation
  • Profound effects on cell proliferation at 5-20 μM
  • Exhibits high selectivity against other kinases
  • Inhibition is competitive with respect to ATP (Ki=95 nM)
  • For research use only
Catalog No. 50-003-74908