Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethanol, 2,2'-dithiobis-, bis(4-methylbenzenesulfonate) | 69981-39-1 | MFCD28976689 | 99.5% | 462.62 g/mol | C18H22O6S4 | 100 MG
SDP

Catalog No. 5000203973
Encompass_Preferred

Bis-Tos-(2-hydroxyethyl disulfide) is a cleavable disulfide linker used to introduce a disulfide-cleavable spacer in antibody-drug conjugate synthesis. Supplied as a high-purity solid, it is suitable for laboratory conjugation chemistry and requires cold, light-protected storage for long-term stability.

  • High purity suitable for synthesis and analytical work.
  • Molecular weight 462.62 g/mol and formula C18H22O6S4.
  • White to off-white solid appearance.
  • Storage: solid at -20°C; in solvent at -80°C for long-term preservation.
  • Available in small research pack sizes for laboratory use.

Catalog No. 50-002-03973 Supplier Medchemexpress LLC Supplier No. HY140126100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Bis-Tos-(2-hydroxyethyl disulfide) is a cleavable disulfide linker used to introduce a disulfide-cleavable spacer in antibody-drug conjugate synthesis. Supplied as a high-purity solid, it is suitable for laboratory conjugation chemistry and requires cold, light-protected storage for long-term stability.

  • High purity suitable for synthesis and analytical work.
  • Molecular weight 462.62 g/mol and formula C18H22O6S4.
  • White to off-white solid appearance.
  • Storage: solid at -20°C; in solvent at -80°C for long-term preservation.
  • Available in small research pack sizes for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.