Learn More
Medchemexpress LLC Ethanol, 2,2'-[iminobis(2,1-ethanediyloxy)]bis- | 54384-47-3 | 97.0% | 193.24 | C8H19NO4 | 5 G
SDP
Catalog No. 5000203372
Encompass_Preferred
NH-bis-PEG2 is a non-cleavable two-unit polyethylene glycol (PEG) linker used in the synthesis of antibody-drug conjugates (ADCs) and PROTACs. It is supplied as a solid with a reported purity of 97.0% (NMR) and CAS number 54384-47-3. Available pack sizes range from 100 mg to 50 g, including a 5 g package.
  • Non-cleavable two-unit PEG linker
  • Suitable for ADC and PROTAC synthesis
  • Purity 97.0% (NMR)
  • Available in multiple pack sizes including 5 g
  • Analytical data available: COA, HNMR, MS, SDS
Catalog No. 50-002-03372 Supplier Medchemexpress LLC Supplier No. HY1303285G
NH-bis-PEG2 is a non-cleavable two-unit polyethylene glycol (PEG) linker used in the synthesis of antibody-drug conjugates (ADCs) and PROTACs. It is supplied as a solid with a reported purity of 97.0% (NMR) and CAS number 54384-47-3. Available pack sizes range from 100 mg to 50 g, including a 5 g package.
  • Non-cleavable two-unit PEG linker
  • Suitable for ADC and PROTAC synthesis
  • Purity 97.0% (NMR)
  • Available in multiple pack sizes including 5 g
  • Analytical data available: COA, HNMR, MS, SDS