Learn More
Medchemexpress LLC Ethanol, 2,2'-[iminobis(2,1-ethanediyloxy)]bis- | 54384-47-3 | 97.0% | 193.24 g/mol | C8H19NO4 | 10 G
SDP
Supplier:  Medchemexpress LLC HY13032810G
Encompass_Preferred
NH-bis-PEG2 is a non-cleavable, two-unit polyethylene glycol (PEG) linker with terminal primary amine functionality supplied as a solid for research use. It is designed to serve as a short, flexible spacer in the synthesis of antibody-drug conjugates (ADCs) and proteolysis-targeting chimeras (PROTACs), facilitating conjugation between small molecules and biomolecules.
  • Non-cleavable two-unit PEG spacer.
  • Terminal primary amine for conjugation chemistry.
  • Short, flexible linker suitable for ADC and PROTAC synthesis.
  • High purity suitable for research applications.
  • Available in multiple pack sizes, including 10 G.
Catalog No. 50-002-02012