Learn More
Medchemexpress LLC Ethanol, 2,2'-[iminobis(2,1-ethanediyloxy)]bis- | 54384-47-3 | 97.0% | 193.24 g/mol | C8H19NO4 | 25 G
SDP
Catalog No. 5000204121
Encompass_Preferred
NH-bis-PEG2 is a non-cleavable two-unit polyethylene glycol (PEG) linker containing a central secondary amine, supplied for research use in bioconjugation applications such as antibody-drug conjugate and PROTAC synthesis.
  • Two-unit PEG spacer with a central amine.
  • Liquid, colorless to light yellow appearance.
  • Molecular weight 193.24 g/mol and formula C8H19NO4.
  • High purity (97.0%) suitable for research applications.
  • Hydrophilic linker ideal for spacing and solubilization in conjugates.
  • Available in multiple package sizes, including 25 G.
Catalog No. 50-002-04121 Supplier Medchemexpress LLC Supplier No. HY13032825G
NH-bis-PEG2 is a non-cleavable two-unit polyethylene glycol (PEG) linker containing a central secondary amine, supplied for research use in bioconjugation applications such as antibody-drug conjugate and PROTAC synthesis.
  • Two-unit PEG spacer with a central amine.
  • Liquid, colorless to light yellow appearance.
  • Molecular weight 193.24 g/mol and formula C8H19NO4.
  • High purity (97.0%) suitable for research applications.
  • Hydrophilic linker ideal for spacing and solubilization in conjugates.
  • Available in multiple package sizes, including 25 G.