Learn More
Medchemexpress LLC Ethanol, 2,2'-[iminobis(2,1-ethanediyloxy)]bis- | 54384-47-3 | MFCD00239212 | 97.0% | 193.24 g/mol | C8H19NO4 | 1 G
SDP
Catalog No. 5000202479
Encompass_Preferred
NH-bis-PEG2 is a short, non-cleavable two-unit polyethylene glycol (PEG) linker used as a hydrophilic spacer in conjugation chemistry. It is commonly employed in the synthesis of antibody-drug conjugates (ADCs) and proteolysis targeting chimeras (PROTACs) for research applications.
  • Non-cleavable two-unit PEG linker
  • Used for ADC and PROTAC synthesis
  • High purity: 97.0% (NMR)
  • Molecular weight 193.24 g/mol and formula C8H19NO4
  • Available as a 1 G package
  • Store protected from light at 4°C; in solvent, -80°C (up to 6 months) or -20°C (up to 1 month)
Catalog No. 50-002-02479 Supplier Medchemexpress LLC Supplier No. HY1303281G
NH-bis-PEG2 is a short, non-cleavable two-unit polyethylene glycol (PEG) linker used as a hydrophilic spacer in conjugation chemistry. It is commonly employed in the synthesis of antibody-drug conjugates (ADCs) and proteolysis targeting chimeras (PROTACs) for research applications.
  • Non-cleavable two-unit PEG linker
  • Used for ADC and PROTAC synthesis
  • High purity: 97.0% (NMR)
  • Molecular weight 193.24 g/mol and formula C8H19NO4
  • Available as a 1 G package
  • Store protected from light at 4°C; in solvent, -80°C (up to 6 months) or -20°C (up to 1 month)