Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethanol, 2,2'-[iminobis(2,1-ethanediyloxy)]bis- | 54384-47-3 | MFCD00239212 | 97.0% | 193.24 g/mol | C8H19NO4 | 500 MG
SDP

Catalog No. 5000204013
Encompass_Preferred

NH-bis-PEG2 is a bifunctional polyethylene glycol (PEG) linker used in the synthesis of proteolysis-targeting chimeras (PROTACs) and antibody-drug conjugates (ADCs). It is supplied as a colorless to light yellow liquid, with a molecular weight of 193.24 g/mol, a molecular formula of C8H19NO4, and typical purity of 97.0%.

  • Two-unit polyethylene glycol (PEG) linker
  • Non-cleavable linker suitable for ADC and PROTAC synthesis
  • Colorless to light yellow liquid
  • Molecular weight 193.24 g/mol, formula C8H19NO4
  • Purity 97.0%
  • Available in multiple package sizes, including 500 MG

Catalog No. 50-002-04013 Supplier Medchemexpress LLC Supplier No. HY130328500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NH-bis-PEG2 is a bifunctional polyethylene glycol (PEG) linker used in the synthesis of proteolysis-targeting chimeras (PROTACs) and antibody-drug conjugates (ADCs). It is supplied as a colorless to light yellow liquid, with a molecular weight of 193.24 g/mol, a molecular formula of C8H19NO4, and typical purity of 97.0%.

  • Two-unit polyethylene glycol (PEG) linker
  • Non-cleavable linker suitable for ADC and PROTAC synthesis
  • Colorless to light yellow liquid
  • Molecular weight 193.24 g/mol, formula C8H19NO4
  • Purity 97.0%
  • Available in multiple package sizes, including 500 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.