Learn More
Medchemexpress LLC Ethanol, 2,2'-oxybis-, 1,1'-bis(4-methylbenzenesulfonate) | 7460-82-4 | 99.8% | 414.49 | 25 G
SDP
Catalog No. 5000366664
Encompass_Preferred
Diethylene glycol bis(p-toluenesulfonate) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed to induce selective degradation of target proteins by linking two ligands: one that binds to an E3 ubiquitin ligase and another that targets the protein for degradation through the ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs exploit the ubiquitin-proteasome system
  • Facilitates selective degradation of target proteins
Catalog No. 50-003-66664 Supplier Medchemexpress LLC Supplier No. 25GHYW010948
Diethylene glycol bis(p-toluenesulfonate) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed to induce selective degradation of target proteins by linking two ligands: one that binds to an E3 ubiquitin ligase and another that targets the protein for degradation through the ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs exploit the ubiquitin-proteasome system
  • Facilitates selective degradation of target proteins