Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethanol, 2,2'-oxybis-, 1,1'-dimethanesulfonate | 34604-52-9 | 97.0% | C6H14O7S2 | 25 G
SDP

Catalog No. 5000352523
Encompass_Preferred

Ms-PEG2-MS is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs are compounds that contain two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade specific target proteins, offering a promising approach for targeted therapy.

  • Purity of 97.0%
  • Off-white to light yellow solid appearance
  • Molecular weight of 262.30
  • Chemical formula C6H14O7S2
  • Used in the synthesis of PROTACs
  • CAS number 34604-52-9

Catalog No. 50-003-52523 Supplier Medchemexpress LLC Supplier No. 25GHY140382
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ms-PEG2-MS is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs are compounds that contain two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade specific target proteins, offering a promising approach for targeted therapy.

  • Purity of 97.0%
  • Off-white to light yellow solid appearance
  • Molecular weight of 262.30
  • Chemical formula C6H14O7S2
  • Used in the synthesis of PROTACs
  • CAS number 34604-52-9

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.