Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethanol, 2-[[4-[(7-chloro-4-quinolinyl)amino]pentyl)amino]-, ethanedioate (1:1) | 14142-64-4 | 99.63% | 397.85 | 5 MG
SDP

Catalog No. 5000364176
Encompass_Preferred

Cletoquine oxalate is a major active metabolite of Hydroxychloroquine, produced in the liver by CYP2D6, CYP3A4, CYP3A5, and CYP2C8 isoenzymes. This Chloroquine derivative demonstrates activity against the chikungunya virus (CHIKV) and exhibits antimalarial effects, showing potential for autoimmune disease treatment.

  • Major active metabolite of Hydroxychloroquine.
  • Produced by CYP2D6, CYP3A4, CYP3A5, and CYP2C8 isoenzymes.
  • Active against chikungunya virus (CHIKV).
  • Exhibits antimalarial effects.
  • Potential for autoimmune diseases treatment.
  • Purity: 99.63%.
  • Appearance: Light yellow to yellow solid.
  • Recommended storage at 4°C, sealed and away from moisture.

Catalog No. 50-003-64176 Supplier Medchemexpress LLC Supplier No. 5MGHY135810A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cletoquine oxalate is a major active metabolite of Hydroxychloroquine, produced in the liver by CYP2D6, CYP3A4, CYP3A5, and CYP2C8 isoenzymes. This Chloroquine derivative demonstrates activity against the chikungunya virus (CHIKV) and exhibits antimalarial effects, showing potential for autoimmune disease treatment.

  • Major active metabolite of Hydroxychloroquine.
  • Produced by CYP2D6, CYP3A4, CYP3A5, and CYP2C8 isoenzymes.
  • Active against chikungunya virus (CHIKV).
  • Exhibits antimalarial effects.
  • Potential for autoimmune diseases treatment.
  • Purity: 99.63%.
  • Appearance: Light yellow to yellow solid.
  • Recommended storage at 4°C, sealed and away from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.