Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, 200 proof (100%), Tax-Paid, Decon™ Labs
GSA_VAGREENER_CHOICE

Catalog No. 07678007 Shop All Decon Labs Products
Change view
Click to view available options
Quantity:
0.125 gal.
1 gal.
5 gal.
4 x 1 gal.
55 gal.
24 x 0.125 gal.
Packaging:
Drum
Glass Bottle
Metal Container
Plastic Bottle
Poly Bottle
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
07-678-007 0.125 gal. Glass Bottle
04-355-450 0.125 gal. Plastic Bottle
04-355-451 1 gal. Plastic Bottle
07-678-006 24 x 0.125 gal. Glass Bottle
04-355-721 4 x 1 gal. Glass Bottle
04-355-452 5 gal. Plastic Bottle
22-032-104 5 gal. Poly Bottle
04-355-456 55 gal. Drum
04-355-460 55 gal. Metal Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 07-678-007 Supplier Decon Laboratories, Inc. Supplier No. 2716GEATP
 Restricted Item
Add to Cart
Add to Cart

Federal Tax is included in the price. No federal permit required for purchase.

Used for research, healthcare, industrial, or production applications as a cleaner, solvent or reagent.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 64-17-5 , 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07 g/mol
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name Ethanol
SMILES CCO

Specifications

Melting Point -114°C
CAS Min % 99.9%
Color Clear
CAS Max % 100%
Density 6.62
Quantity 0.125 gal.
Packaging Glass Bottle
Physical Form Liquid
UN Number UN1170
Boiling Point 78.3°C
Percent Purity 100%
Grade USP
Chemical Name or Material Ethanol, 200 Proof (100%)
Show More Show Less
DOTInformation DOT Class: 3, Packaging II
Recommended Storage Do not store alcohol above 120°F.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.