Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, 200 proof (100%), USP, Decon™ Labs
GSA_VAGREENER_CHOICE

Catalog No. 07678005 Shop All Decon Labs Products
Change view
Click to view available options
Quantity:
0.125 gal.
5 gal.
4 x 1 gal.
55 gal.
24 x 0.125 gal.
Packaging:
Cubitainer with Spigot
Glass Bottle
Metal Container
Plastic Bottle
Poly Bottle
Poly Bottle with Spigot
Poly Drum
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
07-678-005 0.125 gal. Glass Bottle
07-678-004 24 x 0.125 gal. Glass Bottle
04-355-222 24 x 0.125 gal. Plastic Bottle
04-355-720 4 x 1 gal. Glass Bottle
04-355-223 - Plastic Bottle
22-032-601 5 gal. Cubitainer with Spigot
04-355-233 - Metal Container
04-355-224 - Poly Bottle
22-032-103 - Poly Bottle with Spigot
04-355-230 - Metal Container
04-355-600 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 07-678-005 Supplier Decon Laboratories, Inc. Supplier No. 2716GEA

This product is not available for sale online. Please contact your sales rep or Customer Service at 1-800-766-7000.

missing translation for 'validMainframeMsg'

May require a federal tax exempt certificate.

Used for research, healthcare, industrial, or production applications as a cleaner, solvent or reagent.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07 g/mol
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name Ethanol
SMILES CCO

Specifications

Melting Point -114°C
Color Clear
CAS Min % 99.9%
Density 6.62
Quantity 0.125 gal.
Packaging Glass Bottle
UN Number UN1170
Boiling Point 78.3°C
Percent Purity 100%
Grade BP/EP/FCC/JP/NF/USP
Chemical Name or Material Ethanol, 200 Proof (100%)
Show More Show Less
DOTInformation DOT Class: 3, Packaging II
Recommended Storage Do not store Ethanol at temperature above 120°F
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.