Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, 200 Proof (100%), USP/EP/ACS, Tax-Paid, Fisher Chemical™

Catalog No. a4094 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Glass Bottle
Steel Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A4094 4 L Glass Bottle
A40920 20 L Steel Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A4094
Only null left
Add to Cart

A tax-paid 200 proof ethanol that meets USP, EP, and ACS testing specifications to encourage cross-regional collaboration and potentially reduce laboratory inventory for ethanol applications.

  • Meets USP, EP, and ACS testing specifications
  • Consolidate your purchases into one product code and potentially reduce inventory
  • Suitable for use as a cleaner, solvent, or reagent in various applications
  • Packaged in laboratory-friendly sizes
  • Pricing includes tax
  • Combination of compendial test specifications enables use across regions
  • Intended for research use only; not for human or animal diagnostic purposes
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114°C
Absorbance 0.40 max. at 240nm (USP, EP), 0.30 max at 250-260nm (USP, EP), 0.10 max at 270-340nm (USP, EP)
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.001% max. (ACS)
Water 0.2% max. (ACS)
Boiling Point 78°C
Formula Weight 46.07
Grade ACS/EP/USP
Chemical Name or Material Ethanol, 200 Proof
Show More Show Less
Precautionary Statement
Note: Products are tested to meet the analytical requirements of the noted grade (USP, Ph. EUR or otherwise). Product test results are available on the Certificate of Analysis or upon request for the specific product.
 
Products are for research use or further manufacturing. Products are not for direct administration to humans or animals. It is the responsibility of the final formulator and end-user to qualify, test and/or validate each product for its intended use as an ingredient in the end product and in accordance with applicable regulations (including without limitation WHO-recommended Hand Rub Formulations (Hand Sanitizers) and/or FDA Policy for Preparation of Certain Alcohol-based Hand Sanitizer Products, as updated). Thermo Fisher Scientific expressly disclaims all non-specified warranties, expressed or implied, including the implied warranties of merchantability and fitness for a particular use.

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.