Learn More
LAB ALLEY Ethanol 200 Proof (100%) Undenatured Alcohol USP/FCC Food Grade Kosher Halal
SDP
Catalog No. NC3342986
Encompass
Unlike the FCC and USP monograms which regulate based on the purity of the final product Kosher certification is concerned with every part of the processing and sourcing. As most ethanol is fermented and distilled from grain sugar cane or corn without use of animal materials Kosher-compliance is not very difficult to obtain. Beyond the traditional roles of ethanol in food production Kosher food grade ethanol can be used as an alternative solvent in cases where it is impossible to modify the traditional processing to make it Kosher.
Catalog No. NC3342986 Supplier LAB ALLEY Supplier No. EAP2001GAL
Unlike the FCC and USP monograms which regulate based on the purity of the final product Kosher certification is concerned with every part of the processing and sourcing. As most ethanol is fermented and distilled from grain sugar cane or corn without use of animal materials Kosher-compliance is not very difficult to obtain. Beyond the traditional roles of ethanol in food production Kosher food grade ethanol can be used as an alternative solvent in cases where it is impossible to modify the traditional processing to make it Kosher.